Learn More
US Plastic Corporation WELD ON NO16 CLR CEMENT 16OZ
SDP
Supplier:  US Plastic Corporation 43401
Encompass
Very thin methylene chloride and should be applied with a dropper or syringe. Exceptionally fast setting cement for acrylics. Item can not ship via Air freight of any kind.
Catalog No. NC9249098