Learn More
Cliawaived Inc The WELLlife COVID-19 / Influenza A&B Test differentiates between SARS-COV-2, influenza A and influenza B antigens with a single test (25 tests per box)
SDP
Supplier:  Cliawaived Inc WFWV01P0002
Encompass_Preferred
Differentiate between SARS-COV-2, influenza A, and influenza B antigens with a single test using an anterior nasal swab specimen. Results available in 10 minutes allow for testing and treatment decision-making during the same office visit. Offers a broader detection window for differentiation of SARS-COV-2, influenza A, and influenza B antigens within five (5) days of symptom onset (25 Tests tests per box).
Catalog No. NC2941128