Learn More
Novus Biologicals 12-230 kDa Pre-filled Plates
SDP
Supplier:  Novus Biologicals PSPP03
Encompass
Compatible with Jess, Abby and Wes systems using Compass v2.7 or higher. Includes 8 Plates (pre-filled with Separation Matrix, Stacking Matrix, Split Running Buffer 2 and Matrix Removal Buffer).
Catalog No. NC1004767