Learn More
Novus Biologicals 12-230 kDa Separation Module, 200 Capillaries
SDP
Supplier:  Novus Biologicals SMW004
Encompass
Use with a Detection Module. Reagents for 200 data points includes two parts: Separation box with 8x Capillary Cartridges (25 capillaries per cartridge), 8x Wes 12-230 kDa pre-filled Plates (pre-filled with Separation Matrix 2, Stacking Matrix 2, Split Running Buffer 2 and Matrix Removal Buffer), Sample Buffer, Wash Buffer (Store at RT), plus 1x Standard Pack 1 clamshell with 8x DTT and 8x FL master mix with 29 kDa System Control protein. (Store at 4 C).
Catalog No. NC1193547