Learn More
ProteinSimple 66-440 kDa Pre-filled Plates
Supplier:  ProteinSimple PSPP04
Encompass
Compatible with Jess, Abby and Wes systems using Compass v2.7 or higher. Includes 8 Plates (pre-filled with Separation Matrix, Stacking Matrix, Split Running Buffer 3 and Matrix Removal Buffer).
Catalog No. NC1016755