Learn More
World Precision Instrument Westcott Rounded Blades, 14 cm, Angled
SDP
Supplier:  World Precision Instrument 504492
Encompass
Westcott Rounded-Blade, Side-Angled Spring Scissors are popular spring handled scissors that automatically reopen after each cut. Westcott brand spring scissors give you an ergonomic and comfortable grip, reducing hand strain and fatigue during repetitive cutting tasks. Westcott spring scissors are perfect for ambidextrous use, often preferred by left-handed individuals. The tips of these surgical scissors are 21 mm long and angled to the side, and they are available with blunt tips. They are 14 cm (5.6”) long. They are constructed of surgical grade stainless steel and are suitable for autoclaving or chemical sterilization.
Catalog No. 50-253-9359