Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Western Blotting Boxes Medium Rectangle, Clear, 4 ml, 3½ x 2-9/16 x 1"
SDP

Supplier:  Electron Microscopy Sciences 7047503

Encompass_Preferred

No matter what size your Western blot is, there's a box that fits it perfectly. With boxes that are correctly matched to common blot sizes, you can fit more blots on shakers/rockers while keeping reagent and antibody usage to an absolute minimum. Additionally, all corners are formed at a 90 deg angle and the bottom surface is uniformly flat, further reducing consumption of costly reagents and antibodies. Equipped with an easy-to-open hinged lid trhat reduces evaporation. Applications: mini protein gels (Bio-Rad Protean 3 and similar).
Color: Clear
Max volume: 4 ml
Dimensions: 3½ x 2-9/16 x 1"

Catalog No. 50-192-9210


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.