Learn More
LICOR Western Blotting Kit VII RD (TBS/NITRO)
Supplier:  LICOR 92635014
Encompass_Preferred
This kit contains IRDye 800CW Goat anti-Mouse Secondary Antibody (0.1 mg, 1 mg/mL), IRDye 680RD Goat anti-Rabbit Secondary Antibody (0.1 mg, 1 mg/mL), 500 mL Intercept (TBS) Blocking Buffer, and 10 Odyssey Nitrocellulose Membranes (0.22 µm, 7 cm x 8.5 cm)
Catalog No. NC2225465