Learn More
Fritsch Milling And Sizing Inc Wet Sieve Pan 200 mm Stl. Steel
SDP
Supplier:  Fritsch Milling And Sizing Inc 31.1100.03
Encompass
For wet sieving with test sieves 200 mm dia.
The sieve pan made of stainless steel with outlet completes the sieve stack.
Catalog No. 50-153-4171