Learn More
Selleck Chemical LLC WH-4-023 5mg 837422-57-8 KIN001-112, KIN112
SDP
Supplier:  Selleck Chemical LLC S75655MG
Encompass_Preferred
WH-4-023 (KIN001-112, KIN112) is a potent and orally active Lck/Src inhibitor with IC50s of 2 nM and 6 nM in cell-free assays, respectively. Exhibits >300-fold selectivity against p38? and KDR. Also potently inhibits SIK (IC50 values are 10, 22 and 60 nM for SIK 1, 2 and 3 respectively) and displays selectivity over a range of closely related kinases. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-7989