Learn More
Medical Chemical Corporation Gomori's Trichrome, 1 gal
SDP
Supplier:  Medical Chemical Corporation 602A1GAL
Encompass_Preferred
Gomori's trichrome is used to produce permanently stained slides from PVA fixed stool samples. SAF and MIF preserved specimens may also be used but there are alternate staining procedures that give better results with these fixatives.
Catalog No. NC1659756