Learn More
Sigma Aldrich Fine Chemicals Biosciences Wheaton sample vials clear
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z1887191PAK
Encompass
Vials are packaged in corrugated trays with partitions then shrink-wrapped to reduce particulate contamination.Autoclavable borosilicate USP Type 1 glass. Order caps separately.
Catalog No. 50-290-0911