Learn More
Sigma Aldrich Fine Chemicals Biosciences WHEATONR CELLine Multiuse1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences DWKWCL100011EA
Encompass
CELLine Bioreactor flask for high density suspension cell culture.Media compartment with cell growth compartment for antibody and protein production.Use with Hybridoma Insect CHO NSO HEK BHK cell linesDisposable and ready-to-useHigh call density and high product concentrationBulk media chamber allows for longer run times without media refreshing and reduces operation timeDecreases use of consumablesCost-efficient space saving and stackableNo additional equipment required for operation
Catalog No. 50-290-4635