Learn More
Medicus Health Wheeled 45 Quart Specimen Transport Cooler
SDP
Supplier:  Medicus Health 5305M2
Encompass
Maxi-Cool Rotomolded Wheeled 45QT Specimen Transport Cooler (with a Convenient Telescopic Handle), goes beyond the minimum standard days of ice retention but up to 10 days or longer (if pre-cooled, kept in low: temps & UV index). Food grade safe, dry ice compatible and tough enough to endure whatever the medical environment throws your way
Catalog No. NC2818121