Learn More
Medchemexpress LLC HY-11067 5mg Medchemexpress, WHI-P97 CAS:211555-05-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY110675MG
Encompass_Preferred
Medchemexpress, HY-11067 5mg WHI-P97 CAS:211555-05-4 WHI-P97 is a potent and selective JAK-3 inhibitor. WHI-P97 is effective in preventing the development allergic asthma in vivo. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0785