Learn More
Medchemexpress LLC White mineral oil | 8042-47-5 | 25 ML
SDP
Supplier:  Medchemexpress LLC 25MLHY139411
Encompass_Preferred
White mineral oil is a highly refined mineral oil composed of saturated aliphatic and alicyclic nonpolar hydrocarbons. It is biologically and chemically stable, and does not support pathogenic bacterial growth. It can resist moisture, extend, soften, smoothen, and lubricate.
  • Biologically and chemically stable
  • Resists moisture
  • Extends, softens, and smoothens
  • Lubricates
  • Composed of saturated aliphatic and alicyclic nonpolar hydrocarbons
Catalog No. 50-003-49055