Learn More
GA International Removable Solvent-Resistant Color Labels for Containers 2" x 1" / 50.8mm x 25.4mm Rectangle white From -80°C to +100°C (-112°F to 212°F)
Supplier:  GA International AUBR28C33WH
Encompass_Preferred
Solvent and chemical-resistant removable color labels are designed for thermal-transfer barcode printers, offering an exceptional solution for color-coding bottles and similar containers that face exposure to a diverse range of common laboratory reagents and storage in deep-freeze conditions.Designed to withstand the damaging effects of alcohols, xylene, toluene, methyl ethyl ketone (MEK), acetone, thinners, commercial cleaners, window cleaners, and surface sanitizing wipes, these labels prove to be a safeguard against chemical degradation. Their resilience ensures that vital information remains clear, legible, and intact, even when subjected to the most challenging conditions.These freezer-grade labels have a glove-friendly adhesive for effortless application, facilitating easy handling while maintaining a strong and secure surface adhesion. We recommend using our XAR-class of solvent-resistant ribbons for printing to guarantee a chemical-resistant printout that retains its qualit
Catalog No. NC3910705