Learn More
Sino Biological Whitlow 218 linker Antibody (AF 488), Rabbit MAb 100 Tests
Supplier:  Sino Biological 111774R0013D100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, synthetic peptide corresponding to the Whitlow 218 linker, and conjugated with AF 488 under optimum conditions, the unreacted AF 488 was removed.
Catalog No. 50-007-49828