Learn More
Charles River Labs Cell Solutions Whole Blood, 10 mL, Fresh, Kit
Supplier:  Charles River Labs Cell Solutions PB000F2KIT
Encompass
Whole blood is collected from IRB consented healthy human donors in our FDA-registered collection centers. Unless otherwise specified, whole blood is collected in a CPD collection bag and specimen samples are collected in EDTA tubes. We can collect whole blood in any commercially available collection tubes with anticoagulants per your specifications.
Catalog No. 50-225-6206
Only null left
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov