Learn More
Hoefer, Inc Whole Rack Barcode Reader
SDP
Supplier:  Hoefer, Inc STFBR01
Encompass_Preferred
Hoefer Barcode Reader, combing a high-resolution, camera-based imaging system, providing a very effective reading of whole SBS rack tubes. With an integrated sample management software, it is an ideal tool for precise identification of 2D codes and sample traceability, can meet the requirements of sample storage of any lab.Features and Benefits:Decode a whole rack of tubes in less than 1 second, capable of reading whole rack of tubes or a single tube. Direct USB power supply and data transfer via Type C or USB3.1 ports. Use with any 2D-coded tubes in SBS format rack, available for 24, 48, 96 and 240 format SBS racks. Data output choice with Excel, CSV, text, TCP/IP, ODBC, etc. Remote control via TCP/1P, allowing synchronized code-reading in multi-computers. Compact size, suitable for small workbench operation. High resolution camera, to provide clearer pictures and more accurate reads.
Catalog No. NC2956938
Only null left
Add to Cart
Request a Quote