Learn More
Electron Microscopy Sciences Visio-Tek™ Digital Wi-Fi/USB Microscope with High magnification, 500x ~ 600x magnification
SDP
Supplier:  Electron Microscopy Sciences 92108
Encompass_Preferred
Visio-tek digital portable imaging solutions utilize both Wi-Fi and USB transmission and can be connected to iOS and Android smartphones or tablets via the dedicated Visio-tek App, as well as to Windows or Mac computers or laptops through a Wi-Fi/USB connection. Using theVisio-tek free software and application, you can capture, store pictures and video, and take measurements. Flexible, cost effective and user-friendly. Visio-tek microscopes allow Wi-Fi inspections anywhere an image magnification is required. Highest powered model for applications requiring the greatest magnification at close range.
Magnification: 500x ~ 600x
Focus range: 0 ~ 3 mm
Light source: 8 White LEDs
Catalog No. 50-193-0501