Learn More
Electron Microscopy Sciences Visio-Tek™ Digital Wi-Fi/USB Microscope with UV + White Light, 5x ~ 200x magnification
SDP
Supplier:  Electron Microscopy Sciences 92104
Encompass_Preferred
Visio-tek digital portable imaging solutions utilize both Wi-Fi and USB transmission and can be connected to iOS and Android smartphones or tablets via the dedicated Visio-tek App, as well as to Windows or Mac computers or laptops through a Wi-Fi/USB connection. Using theVisio-tek free software and application, you can capture, store pictures and video, and take measurements. Flexible, cost effective and user-friendly. Visio-tek microscopes allow Wi-Fi inspections anywhere an image magnification is required. For applications with chemical structures that react to UV light. 3 seconds to change light source.
Magnification: 5x ~ 200x
Focus range: 8 ~ 200 mm
Light source: 4 White LEDs + 4 UV LEDs
Catalog No. 50-193-0497