Learn More
Selleck Chemical LLC WIKI4 10mg 838818-26-1
SDP
Supplier:  Selleck Chemical LLC S749010MG
Encompass_Preferred
WIKI4 is a novel Tankyrase inhibitor with IC50 of 15 nM for TNKS2, and leads to inhibition of Wnt/beta-catenin signaling. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1319414