Learn More
Cole-Parmer Wildco 62-C50 Polyester Line, 328 ft (100 m), 110 lb max load
Supplier:  Cole-Parmer 0549933
Encompass_Preferred
Use this solid braided line with sampling equipment that weighs less than 75 lb (34 kg).
Catalog No. NC2097851