Learn More
Medchemexpress LLC Wilforgine | 37239-47-7 | 857.81 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN10721MG
Encompass_Preferred
Wilforgine is a bioactive sesquiterpene alkaloid found in Tripterygium wilfordii Hook. F. It has the ability to induce microstructural and ultrastructural changes in the muscles of Mythimna separata larvae. Its proposed sites of action are calcium receptors or channels within the muscular system.
  • Bioactive sesquiterpene alkaloid
  • Induces microstructural and ultrastructural changes in muscles
  • Acts on calcium receptors or channels
  • Purity: 99.67%
Catalog No. 50-004-14838