Learn More
Medchemexpress LLC Wilforgine | 37239-47-7 | 857.81 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYN1072
Encompass_Preferred
Wilforgine is a bioactive sesquiterpene alkaloid found in *Tripterygium wilfordii Hook. F.* It induces microstructural and ultrastructural changes in the muscles of *Mythimna separata* larvae, with its sites of action believed to be calcium receptors or channels in the muscular system. For research use only.
  • Bioactive sesquiterpene alkaloid
  • Derived from *Tripterygium wilfordii Hook. F.*
  • Induces microstructural and ultrastructural changes
  • Acts on muscles of *Mythimna separata* larvae
  • Proposed to target calcium receptors or channels in the muscular system
Catalog No. 50-003-89376