Learn More
Medchemexpress LLC Wilforgine | 37239-47-7 | 857.81 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN10725MG
Encompass_Preferred
Wilforgine is a bioactive sesquiterpene alkaloid found in *Tripterygium wilfordii Hook. F.*. It induces microstructural and ultrastructural changes in the muscles of *Mythimna separata* larvae, with its proposed sites of action being calcium receptors or channels within the muscular system.
  • Bioactive sesquiterpene alkaloid
  • Found in *Tripterygium wilfordii Hook. F.*
  • Induces microstructural and ultrastructural changes in *Mythimna separata* larvae muscles
  • Targets calcium receptors or channels
Catalog No. 50-004-15087