Learn More
NEOGEN Wilkins-Chalgren Broth, 500 g
SDP
Supplier:  NEOGEN NCM0219A
Encompass_Preferred
Wilkins-Chalgren Broth is used for the cultivation of anaerobic microorganisms in a laboratory setting. Wilkins-Chalgren Broth is not intended for use in the diagnosis of disease or other conditions in humans. Wilkins-Chalgren Broth is derived from Wilkins-Chalgren Agar. Wilkins-Chalgren Broth provides good growth of anaerobes. This medium was selected because it does not require the addition of blood to support satisfactory growth of most anaerobes. Wilkins-Chalgren Broth is the same formula as Wilkins-Chalgren Agar with agar omitted. The survival of anaerobic bacteria is dependent on their sensitivity to oxygen, nutritional requirements, appropriate collection, culture medium, and incubation time and temperature.For more information visit https://www.neogen.com/solutions/Microbiology/wilkins-chalgren-broth
Catalog No. 50-201-5765