Learn More
Cambridge Isotope Laboratories Wilmad Ultra-High Field Microprobe 5 Mm 8"""" 500 Mhz 1 Mm O.D. Nmr Tube, 1 Tube
Supplier:  Cambridge Isotope Laboratories 6201A
Encompass
This item has a minimum order quantity of 2 per supplier requirements
Wilmad Ultra-High Field Microprobe 5 Mm 8"""" 500 Mhz 1 Mm O.D. Nmr Tube, 1 Tube
Catalog No. 50-162-9032