Learn More
Enzo Life Sciences WIN-55,212-2 (10mg). CAS: 131543-23-2
SDP
Supplier:  Enzo Life Sciences BMLCR1050010
Encompass_Preferred
High affinity cannabinoid receptor (CB) agonist (Ki=62.3nM and 3.3nM at the human cloned CB1 and CB2 receptors, respectively). Potent analgesic in a rat model of neuropathic pain. Activates p42 and p44 MAP kinase via a receptor-mediated signaling. Alternative name: (R)-(+)-[2,3-Dihydro-5-methyl-3-(4-morpholinylmethyl)pyrrolo[1,2,3-de]-1,4-benzoxazin-6-yl]-1-naphthalenylmethanone . mesylate. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO; insoluble in water. Long Term Storage: +4°C.
Catalog No. 50-201-0467