Learn More
Medchemexpress LLC 1-methylimidazole-d6 | 285978-27-0 | MFCD00190485 | 98.8% | 88.14 | C4D6N2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY59137S10MG
Encompass_Preferred
1-Methylimidazole-d6 is the deuterium-labeled isotopologue of 1-methylimidazole supplied for use as an isotopic reference and internal standard in analytical laboratories. It is intended for tracer and quantitative applications in NMR, GC-MS, and LC-MS workflows.
  • Deuterium-labeled internal standard for quantitative analysis by NMR, GC-MS, and LC-MS.
  • High chemical purity (≈98.8%) and consistent isotopic enrichment for reliable results.
  • Molecular weight 88.14 and molecular formula C4D6N2 for precise mass calculations.
  • Available in small, laboratory-scale quantities suitable for standard analytical protocols.
Catalog No. 50-002-98730