Learn More
Chemplex Industries WINDOW FRAME 0.14 MIL 2.5IN DI
Supplier:  Chemplex Industries 3013
Encompass_Preferred
CHEMPLEX SPECTROMEMBRANE(TM) Thin-Film Sample Support Window Frames, Mylar(TM), 3.6 u (0.14 mil), 2.5 Dia., In.
Catalog No. 50-249-350