Learn More
Grainger Window Screen Pointer Latchs Black PK20
Supplier:  Grainger 170F61
Encompass_Preferred
The Grainger 170F61 is a 1/4 HP, 115V, 1-phase, 60Hz, 3450 RPM, 56C frame, open drip proof, ball bearing, C-face, electric motor. It has a NEMA 48 frame size with a 1.25" shaft diameter and 1.38" shaft height. The motor is totally enclosed, fan cooled (TEFC) with a cast iron body, aluminum end brackets, and Class F insulation. It is designed for general purpose applications.
Catalog No. 50-276-5832