Learn More
Essendant WIPEA-PURPJ9.8X12.2BE
Supplier:  Essendant KCC33241
Encompass_Preferred
If you have a task where wiping precision and reliability are important WypAll Oil Grease and Ink Cloths are an excellent choice They are designed for wiping tasks where low lint or the use of chemicals is required as the polypropylene sheets are specially engineered for critical applications involving solvents inks and paints These absorbent sheets are acid- base- and solvent-resistant and ideal for use with lubricants oil and grease Whether you re responsible for cleaning by hand manufacturing/assembly pre-packaging wipe-down or refinishing these industrial wipers are essential tools of the trade Jumbo Roll Towel/Wipe Type- Shop Towels Applicable Material- Multiple Material(s)- Polypropylene Number of Plies- 1
Catalog No. 50-000-32728