Learn More
Essendant Chem-Ready Dry Wipes, 1-Ply, 5 x 2.16, Unscented, White, 180/Roll, 6 Rolls/Carton
Supplier:  Essendant TRK192807
Encompass_Preferred
Everwipe Chem-Ready material is specifically designed to work with all the most popular types of sanitizers and disinfectants. This exclusive material performs like a wiper but does not bind quaternary chemicals like ordinary towels and wipers can. The unique material apertures not only trap particles they help scrub the surface. Delivery system allows you to create your own wet wipe system with whatever surfactant is best for your application. The closed system greatly reduces the possibility of surface cross-contamination of pathogens. The small roll put-up when used in the Chem-Ready bucket is extremely portable, easily resealed and spill proof. Ideal for use with quaternary, bleach and phenol-based compounds. The system is cleaner, safer and easier than open bucket or spray type systems.
Catalog No. 26-780-40