Learn More
Essendant WIPERQTR FLD500/CSBE
Supplier:  Essendant HOSNA115QPB
Encompass_Preferred
Advanced performance wiper is creped spunlace which is very low lint and highly absorbent It s cloth-like feel resists solvents and is ideal for automotive and aerospace prep and cleaning A great shop towel and rag replacement Towel/Wipe Type- Dry Wipes Applicable Material- Equipment Machines Material(s)- Spunlace Color(s)- Blue
Catalog No. 50-000-34819