Learn More
Essendant WIPESCRTCL TSK12X15400
Supplier:  Essendant KCC06121
Encompass_Preferred
Kimtech Prep SCOTTPURE Critical Task Wipers have excellent absorbency and are really low-lint The special rayon/polyester spunlace material is solvent-resistant and have no binders or surfactants These white wipers work well in aerospace clean manufacturing areas and DVD making and printing They are designed for critical tasks where precision and reliability are important Quarterfold Towel/Wipe Type- Folded Wipers Fold Types- Quarter-Fold Applicable Material- Multiple Material(s)- Rayon/Polyester
Catalog No. 50-000-31664