Learn More
Essendant WIPESCRTCL TSK7X76/225
Supplier:  Essendant KCC06193
Encompass_Preferred
Kimtech Prep SCOTTPURE Critical Task Wipers have excellent absorbency and are really low-lint The special rayon/polyester spunlace material is solvent-resistant and have no binders or surfactants These white wipers work well in aerospace clean manufacturing areas and DVD making and printing They are designed for critical tasks where precision and reliability are important Center-Pull Towel/Wipe Type- Center-Pull Towels Applicable Material- Multiple Material(s)- Rayon/Polyester Number of Plies- 1
Catalog No. 50-000-33509