Learn More
Essendant WIPESRAG RPLCMBE6/130
Supplier:  Essendant KCC35431
Encompass_Preferred
WypAll General Clean X60 Cloths absorb more water than rags They bring the absorbing power of HydroKnit technology (for superior absorbency) to a lighter-weight cloth Designed to outperform laundered rags the clothes help raise the standard for general-purpose cloths at a terrific value They don t use adhesives or binders so they ll leave no residue behind and they re reinforced for strength when wet or dry These cleaning cloths are so sturdy that they can often be used more than once (once you ve used them a few times they re disposable) They re designed to reduce waste so you spend less Small Roll Towel/Wipe Type- Shop Towels Material(s)- Hydroknit Color(s)- Blue Scent- Unscented
Catalog No. 50-000-33298