Learn More
Essendant WIPESWYPAL X9011.1X16.8
Supplier:  Essendant KCC12891
Encompass_Preferred
When you have a tough job reach for WypAll X90 Cloths with their innovative design they have the highest fluid capacity of any WypAll Brand product they can lift and release fluid In fact they provide 75pct more oil absorption and 35pct more water absorption than Wypall X80 Cloth They deliver superb fluid handling which allows you to accomplish clean wiping applications like never before They re ideal for solvent wiping surface preparation equipment cleaning repair and maintenance tasks so when you need durability cleanliness and high fluid capacity reach for WypAll X90 Cloths Cloths contain no glues adhesives or binders which means there are low extractables for very clean solvent wiping They feature outstanding fluid reservoir capacity for absorbing or delivering fluids so you ll waste less of your valuable solvents (they release 90pct of the solvents they absorb outperforming rags and reducing your solvent spend) In addition they feature increased durability
Catalog No. 50-000-31580