Learn More
Essendant WIPESWYPALL L309.8X15.2
Supplier:  Essendant KCC05820
Encompass_Preferred
Regardless of your task industry or budget our WypAll wipes towels and cloths are designed to give you the durability absorbency and versatility of a laundered cloth with the cost efficiency and hygiene of a disposable When you re looking for a lighter-weight towel at an everyday great price you can trust WypAll L30 Towels They are designed with the DRC (double re-creping) process which gives you high surface softness (a cloth-like feel) with a reinforced grid that s strong for wet surface wiping and cleaning These WypAll L30 Towels are a great choice to replace air laid towels They re soft enough to use on skin so they re popular for use in healthcare and patient care They re also tough enough for light-duty processing janitorial tasks and maintenance (as an alternative to cloth rags) These towels are a trusted tool to help you get the job done right As we look towards the future Kimberly-Clark Professional is committed to providing a wide collection of essential produ
Catalog No. 50-000-34003