Learn More
Essendant WIPESWYPALL L40POPUP
Supplier:  Essendant KCC05740
Encompass_Preferred
Regardless of your task industry or budget our WypAll wipes towels and cloths are designed to give you the durability absorbency and versatility of a laundered cloth with the cost efficiency and hygiene of a disposable Trust the WypAll Brand the one that sets the standard for excellence in wipers WypAll L40 Towels are effective wipers that absorb liquids lightning-fast These towels are made of bonded cellulose using DRC (double re-creped) technology for strength softness and absorbency As a wiping towel it soaks up liquids fast but all you ll feel is softness thanks to a plush touchable outer layer In fact it s a popular choice for patient wiping in healthcare industries for bathing and patient drying These utility towels are specially designed to absorb liquids lubricants and oils (in restaurants for wiping counters in mechanic shops industrial and commercial factories and more) yet they re soft enough for use on face and hands Experience the difference in super
Catalog No. 50-000-33272