Learn More
Essendant WIPESWYPALL REFILLSWE
Supplier:  Essendant KCC83571
Encompass_Preferred
WypAll is the number one brand of disposable wipes in the US and Canada Maximize performance while minimizing time effort and product waste If you ve been using either laundered shop towels or textile shop rags it s time to make the switch to WypAll PowerClean X70 Cloths These medium-duty cloths are durable absorbent industrial cloths featuring high-tech Hydroknit material WypAll X70 cloths now absorb up to 20pct more oil and water than before In addition they feature a new cloth-like look and feel for enhanced absorbency making them even more effective for your cleaning tasks These cloths are made of soft pulp fibers bonded to a polypropylene base sheet for absorbency and tear resistance making them suitable for removing dirt oil grime and solvents in a variety of industrial and manufacturing industries These cloths are so sturdy that they can often be used more than once (Once you ve used them a few times they re disposable ) These premium industrial cleaning clo
Catalog No. 50-000-33304