Learn More
Essendant WIPESWYPALL X80BE
Supplier:  Essendant KCC41041
Encompass_Preferred
WypAll is the number one brand of disposable wipes in the US and Canada Maximize performance while minimizing time effort and product waste If you ve been using industrial cloth towels it s time to make the switch to WypAll PowerClean X80 Cloths These cloths feature innovations like high-tech Hydroknit fast-absorbing material and are durable for scrubbing WypAll X80 cloths now absorb over 20pct more oil and up to 15pct more water than before In addition they now feature a tough precision-engineered surface texture making them even more effective for your cleaning tasks The heavy-duty cloths are made of soft pulp fibers bonded to a polypropylene base sheet for absorbency and durability making them suitable for removing dirt oil grime and solvents in industrial and manufacturing businesses The cloths are reusable and reinforced for strength when wet and dry These bulk cloths are a great alternative safer and more efficient to replace laundered rental shop towels a
Catalog No. 50-000-33194