Learn More
Quantum Storage Systems WIRE BASKET DIVIDER 14X9
Supplier:  Quantum Storage Systems 149DC
Encompass_Preferred
Wire Basket Divider, Chrome Finish, 14" Length x 9" Height Product features • All welded construction with additional wire trussing for high strength characteristics • Wire allows air to circulate and light to penetrate for increased product visibility • 600 to 800 lbs capacity per shelf • Minimal dirt accumulation • This is divider without basket, optional dividers can be used to separate product
Catalog No. 50-752-0552