Learn More
MIDWEST WHOLESALE CONTAINER WIRE MESH EXTENDED BOX TRUCK
Supplier:  MIDWEST WHOLESALE CONTAINER SS3636EB
Encompass_Preferred
36 x 36 x 48 OAH SS wire mesh expanded box truck: 1 x 1 x 3/16 angle, .75 x no 11 expanded flat mesh, .375 rod handle, SS washdown casters.T304 SS #4 finish
Catalog No. NC2915828
Add to Cart
Request a Quote