Learn More
Grainger Wire Rope Locking Stud LH Thread 7/32 in
Supplier:  Grainger 16X730
Encompass_Preferred
The Grainger 16X730 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is thermally protected and rated for continuous duty applications. It features a cast iron body with standard mounting feet and comes pre-wired with color coded leads. This ODP motor would be well suited for driving pumps, fans, conveyors, compressors, machine tools, or other industrial equipment. With proper installation and maintenance, this motor should provide reliable operation.
Catalog No. 50-276-7089