Learn More
Grainger Wire Rope Locking Stud LH Thread 5/32 in
Supplier:  Grainger 16X722
Encompass_Preferred
The Locoloc Wire Rope Stud is a 5/32 inch wire rope stud that is compatible with 5/32 inch wire rope. It is made from zinc plated steel and has a working load limit of 920 lbs. The stud has a head diameter of 0.28 inches and an overall length of 0.63 inches. It can be used for creating an endless wire rope sling and features a collar stop to hold it in place. This wire rope stud is ideal for industrial lifting applications.
Catalog No. 50-276-7079