Learn More
Grainger Wire Rope Locking Stud RH Thread 3/32 in
Supplier:  Grainger 16X713
Encompass_Preferred
The Locoloc Wire Rope Stud 3/32 in Compatible is a wire rope stud designed for use with 3/32 inch diameter wire rope. It is made from zinc plated steel and has a working load limit of 110 lbs. The stud has a head diameter of 0.28 inches and an overall length of 0.5 inches. It can be used for creating terminations and connections in wire rope assemblies. This wire rope stud is compatible with Locoloc fittings and accessories. It provides a secure termination point for small diameter wire ropes used in a variety of applications.
Catalog No. 50-276-8194