Learn More
Grainger Wire Rope Oval Sleeve 1/16 in Copper
Supplier:  Grainger 16X776
Encompass_Preferred
The LOOS Wire Rope Oval Sleeve is an oval-shaped wire rope sleeve made of drop forged steel with a zinc finish. It has an inside diameter of 16mm and is compatible with 6x7 and 6x19 wire rope constructions. This wire rope sleeve can be used for various rigging and lifting applications to help form eyes at the ends of wire rope for attaching hooks, shackles and other hardware. The oval shape provides greater holding power than round sleeves. This wire rope sleeve meets Fed Spec RR-S-550 and is rated for 1770 lb working load limit.
Catalog No. 50-276-8039